Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201642.4 | NP_964004.1 | 1 | 280 | ||
GenBank mRNA | AK128918.1 | - | 1 | 206 | ||
AK144599.1 | - | 1 | 172 | |||
AK153157.1 | - | 1 | 170 | |||
AK154973.1 | - | 1 | 204 | |||
AK159496.1 | - | 1 | 218 | |||
AK162317.1 | - | 1 | 204 | |||
AK165465.1 | - | 1 | 204 | |||
BC141205.1 | - | 1 | 189 | 9055817 | ||
BC141206.1 | - | 1 | 189 | 9055818 |