Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msantd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207277.1 | NP_997160.1 | 1, 2 | 1990 | ||
GenBank mRNA | AK044284.1 | - | 1, 2 | 1990 | ||
BC116928.1 | - | 1, 2 | 1058 | 40126247 | ||
BC116930.1 | - | 1, 2 | 1058 | 40126249 |