Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011151.2 | NP_035281.1 | 6 | 1559 | ||
GenBank mRNA | AJ271836.1 | - | 6 | 1545 | ||
AK163283.1 | - | 6 | 1552 | |||
D17411.1 | - | 6 | 1184 |