Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048176.1 | NP_001041641.1 | 12 | 1551 | ||
GenBank mRNA | BC046474.1 | - | 12 | 1657 | ||
CJ235603.1 | - | 6 | 482 | |||
HQ426680.1 | - | 12 | 1551 | |||
HQ426681.1 | - | 10 | 1344 | |||
HQ426682.1 | - | 9 | 1212 | |||
HQ426683.1 | - | 8 | 969 | |||
HQ426684.1 | - | 11 | 1419 | |||
HQ426685.1 | - | 11 | 1491 | |||
HQ426686.1 | - | 11 | 1408 | |||
HQ426687.1 | - | 12 | 1682 | |||
HQ426688.1 | - | 12 | 1698 | |||
HQ426689.1 | - | 11 | 1566 | |||
HQ426690.1 | - | 12 | 1768 | |||
HQ426691.1 | - | 1350 | ||||
HQ426692.1 | - | 1481 | ||||
HQ426693.1 | - | 1391 | ||||
HQ426694.1 | - | 1304 | ||||
HQ426695.1 | - | 1523 | ||||
HQ426696.1 | - | 1463 | ||||
HQ426697.1 | - | 1608 | ||||
HQ426698.1 | - | 1316 | ||||
HQ426699.1 | - | 1380 | ||||
HQ426700.1 | - | 1654 | ||||
HQ426701.1 | - | 1670 | ||||
HQ426702.1 | - | 11 | 1133 | |||
HQ426703.1 | - | 9 | 926 | |||
HQ426704.1 | - | 12 | 1290 | |||
HQ426705.1 | - | 10 | 990 | |||
HQ426706.1 | - | 10 | 1077 | |||
HQ426707.1 | - | 10 | 1224 | |||
HQ426708.1 | - | 6 | 587 | |||
HQ426709.1 | - | 9 | 982 | |||
HQ426710.1 | - | 8 | 922 | |||
HQ426711.1 | - | 9 | 1129 |