Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048176.1 | NP_001041641.1 | 12 | 1514 | ||
GenBank mRNA | BC046474.1 | - | 12 | 1620 | ||
CJ235603.1 | - | 6 | 445 | |||
HQ426680.1 | - | 12 | 1514 | |||
HQ426681.1 | - | 10 | 1307 | |||
HQ426682.1 | - | 9 | 1175 | |||
HQ426683.1 | - | 8 | 932 | |||
HQ426684.1 | - | 11 | 1382 | |||
HQ426685.1 | - | 11 | 1454 | |||
HQ426686.1 | - | 11 | 1371 | |||
HQ426687.1 | - | 12 | 1645 | |||
HQ426688.1 | - | 12 | 1661 | |||
HQ426689.1 | - | 11 | 1529 | |||
HQ426690.1 | - | 12 | 1731 | |||
HQ426691.1 | - | 1313 | ||||
HQ426692.1 | - | 1444 | ||||
HQ426693.1 | - | 1354 | ||||
HQ426694.1 | - | 1267 | ||||
HQ426695.1 | - | 1486 | ||||
HQ426696.1 | - | 1426 | ||||
HQ426697.1 | - | 1571 | ||||
HQ426698.1 | - | 1279 | ||||
HQ426699.1 | - | 1343 | ||||
HQ426700.1 | - | 1617 | ||||
HQ426701.1 | - | 1633 | ||||
HQ426702.1 | - | 11 | 1096 | |||
HQ426703.1 | - | 9 | 889 | |||
HQ426704.1 | - | 12 | 1253 | |||
HQ426705.1 | - | 10 | 953 | |||
HQ426706.1 | - | 10 | 1040 | |||
HQ426707.1 | - | 10 | 1187 | |||
HQ426708.1 | - | 6 | 550 | |||
HQ426709.1 | - | 9 | 945 | |||
HQ426710.1 | - | 8 | 885 | |||
HQ426711.1 | - | 9 | 1092 |