Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048176.1 | NP_001041641.1 | 1 | 46 | ||
GenBank mRNA | BC046474.1 | - | 1 | 12 | ||
BY742285.1 | - | 1 | 56 | |||
HQ426680.1 | - | 1 | 46 | |||
HQ426681.1 | - | 1 | 46 | |||
HQ426682.1 | - | 1 | 46 | |||
HQ426683.1 | - | 1 | 46 | |||
HQ426684.1 | - | 1 | 46 | |||
HQ426685.1 | - | 1 | 46 | |||
HQ426686.1 | - | 1 | 46 | |||
HQ426687.1 | - | 1 | 46 | |||
HQ426688.1 | - | 1 | 46 | |||
HQ426689.1 | - | 1 | 46 | |||
HQ426690.1 | - | 1 | 46 |