Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rergl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128090.1 | NP_001121562.1 | 4 | 269 | ||
GenBank mRNA | BC147153.1 | - | 4 | 269 | 9056190 | |
BC147154.1 | - | 4 | 269 | 9056193 | ||
BC147732.1 | - | 4 | 334 | 9056681 | ||
BC147735.1 | - | 4 | 344 | 9056684 |