Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010840.2 | NP_001010840.1 | 1 | 1042 | ||
GenBank mRNA | BC139121.1 | - | 1 | 1042 | 8862143 | |
BC139122.1 | - | 1 | 1042 | 8862144 |
Set Membership: |
![]() |