Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015827.2 | NP_056642.1 | 14, 15 | 1765 | ||
GenBank mRNA | AF043120.1 | - | 3, 4 | 310 | ||
AK088064.1 | - | 14, 15 | 1766 | |||
AK153352.1 | - | 14, 15 | 1766 | |||
BC006675.1 | - | 14, 15 | 1752 | 3499585 | ||
BC010842.1 | - | 14, 15 | 1754 |