Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110147.1 | NP_001103617.1 | 9 | 1698 | ||
NM_016788.3 | NP_058068.2 | 9 | 1698 | |||
GenBank mRNA | AF037260.1 | - | 9 | 1481 | ||
AK087965.1 | - | 6 | 855 | |||
BC052421.1 | - | 9 | 1698 | 6400209 | ||
DQ666696.1 | - | 9 | 1509 |