Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145528.3 | NP_663503.1 | 3 | 444 | ||
GenBank mRNA | AK149672.1 | - | 3 | 403 | ||
AK154470.1 | - | 3 | 430 | |||
AK163962.1 | - | 3 | 396 | |||
AK171206.1 | - | 3 | 430 | |||
AK171440.1 | - | 3 | 421 | |||
BC016669.1 | - | 3 | 428 | 2648797 | ||
BC023673.1 | - | 3 | 394 | 5343236 | ||
BC023712.1 | - | 3 | 417 | 5346303 | ||
BC033419.1 | - | 3 | 395 | 5359944 |