Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177470.3 | NP_803421.1 | 5 | 496 | ||
GenBank mRNA | AK002555.1 | - | 5 | 486 | ||
AK050101.1 | - | 5 | 498 | |||
AK146007.1 | - | 5 | 511 | |||
AK167567.1 | - | 5 | 497 | |||
AK167715.1 | - | 5 | 494 | |||
AK169359.1 | - | 5 | 497 | |||
BC028901.1 | - | 5 | 491 | 4195488 |