Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023175.1 | NP_075664.1 | 7, 8 | 637 | ||
GenBank mRNA | AF284573.1 | - | 7, 8 | 637 | ||
AK003604.1 | - | 4, 5 | 332 | |||
AK004533.1 | - | 7, 8 | 621 | |||
AK004535.1 | - | 7, 8 | 623 | |||
BC020153.1 | - | 7, 8 | 626 | 3992930 | ||
BC080749.1 | - | 1, 2 | 711 |