Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029148.1 | NP_083424.1 | 1 | 267 | ||
GenBank mRNA | AK015667.1 | - | 1 | 279 | ||
AK030696.1 | - | 1 | 281 | |||
AK033111.1 | - | 1 | 189 | |||
AK040779.1 | - | 1 | 251 | |||
AK129301.1 | - | 1 | 266 | |||
AK147465.1 | - | 1 | 268 | |||
BC050918.1 | - | 1 | 166 | |||
BC054558.1 | - | 1 | 267 | 6405599 | ||
BC138168.1 | - | 1 | 241 | 8861187 | ||
BC138169.1 | - | 1 | 241 | 8861188 |