Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF534114.1 | - | 6 | 661 | ||
AK018166.1 | - | 6 | 1470 | |||
AK019847.1 | - | 3 | 5336 | |||
AK086301.1 | - | 7 | 871 | |||
AK134581.1 | - | 7 | 715 | |||
AK153400.1 | - | 7 | 699 | |||
AK184479.1 | - | 1 | 42 | |||
BC005640.1 | - | 7 | 700 | |||
BC018178.1 | - | 7 | 686 | |||
BC037451.1 | - | 6 | 690 | 4018648 | ||
U27462.1 | - | 7 | 908 |