Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016771.3 | NP_058051.3 | 8 | 1092 | ||
GenBank mRNA | AF026073.1 | - | 8 | 970 | ||
AK017162.1 | - | 2 | 286 | |||
AK133530.1 | - | 8 | 1011 | |||
BC010752.1 | - | 8 | 1066 | |||
BC066190.1 | - | 8 | 1045 | 30472198 |