Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016784.3 | NP_058064.2 | 9 | 873 | ||
GenBank mRNA | AF044334.1 | - | 8 | 765 | ||
AK075993.1 | - | 9 | 861 | |||
AK076007.1 | - | 9 | 851 | |||
AK077197.1 | - | 9 | 874 | |||
AK080068.1 | - | 9 | 880 | |||
BC006750.1 | - | 9 | 843 | 3709134 | ||
BC052895.1 | - | 9 | 831 |