Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc40a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016917.2 | NP_058613.2 | 3 | 570 | ||
GenBank mRNA | AF215637.1 | - | 3 | 523 | ||
AF226613.1 | - | 3 | 536 | |||
AF231120.1 | - | 3 | 509 | |||
AK008700.1 | - | 3 | 374 | |||
AK033987.1 | - | 3 | 586 | |||
AK144780.1 | - | 3 | 557 | |||
AK147137.1 | - | 3 | 556 | |||
AK159294.1 | - | 3 | 559 | |||
AK159855.1 | - | 3 | 556 | |||
AK167608.1 | - | 3 | 559 | |||
BC003438.1 | - | 3 | 539 | 2647365 |