Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc40a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016917.2 | NP_058613.2 | 7 | 1124 | ||
GenBank mRNA | AF215637.1 | - | 7 | 1077 | ||
AF226613.1 | - | 7 | 1090 | |||
AF231120.1 | - | 7 | 1063 | |||
AK008700.1 | - | 7 | 928 | |||
AK032732.1 | - | 2 | 118 | |||
AK144780.1 | - | 7 | 1111 | |||
AK147137.1 | - | 7 | 1110 | |||
AK159294.1 | - | 7 | 1113 | |||
AK159855.1 | - | 7 | 1110 | |||
AK167608.1 | - | 7 | 1113 | |||
BC003438.1 | - | 7 | 1093 | 2647365 |