Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016867.1 | NP_058563.1 | 6 | 904 | ||
GenBank mRNA | AF061262.1 | - | 6 | 904 | ||
AK008307.1 | - | 6 | 721 | |||
AK008626.1 | - | 6 | 917 | |||
AK168560.1 | - | 6 | 915 | |||
BC013440.1 | - | 6 | 958 | 4238716 |