Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016843.3 | NP_058539.2 | 5, 6 | 728 | ||
GenBank mRNA | AK002504.1 | - | 5, 6 | 837 | ||
AK003530.1 | - | 5, 6 | 655 | |||
AK047170.1 | - | 5, 6 | 778 | |||
AK049706.1 | - | 5, 6 | 772 | |||
AK077738.1 | - | 5, 6 | 730 | |||
AK078459.1 | - | 5, 6 | 808 | |||
AK153271.1 | - | 5, 6 | 730 | |||
AK161950.1 | - | 745 | ||||
AK165941.1 | - | 5, 6 | 753 | |||
AK166899.1 | - | 5, 6 | 813 | |||
AK170507.1 | - | 5, 6 | 745 | |||
AK170751.1 | - | 5, 6 | 754 | |||
BC016410.1 | - | 5, 6 | 738 | 3968016 | ||
BC046802.1 | - | 5, 6 | 864 | 6404607 | ||
X61506.1 | - | 5, 6 | 842 |