Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019439.3 | NP_062312.3 | 7 | 1008 | ||
GenBank mRNA | AF008649.1 | - | 7 | 892 | ||
AF114168.2 | - | 7 | 906 | |||
AF120255.1 | - | 2 | 336 | |||
AK038844.1 | - | 48 | ||||
AK156397.1 | - | 6 | 893 | |||
BC054735.1 | - | 7 | 1008 | 6401243 | ||
BC056990.1 | - | 2 | 385 | 6821442 |
Set Membership: |
G-protein coupled receptor |