Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018821.4 | NP_061291.2 | 3 | 1300 | ||
GenBank mRNA | AF121907.1 | - | 2 | 1207 | ||
AK019993.1 | - | 4 | 1454 | |||
AK028665.1 | - | 3 | 1294 | |||
AK028701.1 | - | 3 | 1286 | |||
AK031164.1 | - | 3 | 1301 | |||
AK044946.1 | - | 3 | 1296 | |||
AK083341.1 | - | 3 | 1285 | |||
AK142069.1 | - | 3 | 1416 | |||
AK156591.1 | - | 4 | 1507 | |||
BC017597.1 | - | 2 | 1202 | 4482828 | ||
BC085245.1 | - | 4 | 1128 | 30444479 | ||
BC094443.1 | - | 4 | 1355 | 4017254 |