Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173036.3 | NP_766624.3 | 9, 10 | 1233 | ||
GenBank mRNA | AK037149.1 | - | 5, 6 | 2178 | ||
AK154648.1 | - | 9, 10 | 1209 | |||
AK155164.1 | - | 9, 10 | 1205 | |||
BC026426.1 | - | 7, 8 | 939 | |||
BC064726.1 | - | 9, 10 | 1193 | 30433392 | ||
BC071188.1 | - | 7, 8 | 1169 |
Set Membership: |
![]() |