Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019481.2 | NP_062354.2 | 15 | 1744 | ||
GenBank mRNA | AF199365.1 | - | 15 | 1744 | ||
AF199366.1 | - | 15 | 1744 | |||
AK005153.1 | - | 9 | 1078 | |||
AK128899.1 | - | 15 | 1744 | |||
BC022672.1 | - | 11 | 1123 | |||
BC040789.1 | - | 15 | 1718 | |||
BC049981.1 | - | 15 | 1726 | 5065026 |