Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018864.5 | NP_061352.2 | 2, 3 | 189 | ||
GenBank mRNA | AF042730.1 | - | 2, 3 | 134 | ||
AK011770.1 | - | 2, 3 | 127 | |||
AK078406.1 | - | 2, 3 | 138 | |||
AK083238.1 | - | 2, 3 | 123 | |||
AK145472.1 | - | 126 | ||||
AK147778.1 | - | 2, 3 | 149 | |||
AK152774.1 | - | 2, 3 | 140 | |||
AK153457.1 | - | 2, 3 | 156 | |||
AK160203.1 | - | 2, 3 | 127 | |||
AK160467.1 | - | 2, 3 | 121 | |||
AK165977.1 | - | 2, 3 | 131 | |||
AK168457.1 | - | 2, 3 | 164 | |||
BC049080.1 | - | 2, 3 | 130 | 6419835 | ||
BC055722.1 | - | 1, 2 | 84 | 6413389 | ||
BY283562.1 | - | 2, 3 | 190 |