Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018889.3 | NP_061377.2 | 12 | 1677 | ||
GenBank mRNA | AK153819.1 | - | 12 | 1594 | ||
BC052658.1 | - | 12 | 1637 | 30048340 | ||
D84436.1 | - | 12 | 1677 |
Gene Symbol: Ccpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286545.1 | NP_001273474.1 | 10 | 2810 | ||
GenBank mRNA | AK134675.1 | - | 10 | 2707 | ||
BC037183.1 | - | 2 | 779 |