Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019501.3 | NP_062374.2 | 10, 11 | 1188 | ||
GenBank mRNA | AB210841.1 | - | 10, 11 | 1108 | ||
AF118855.1 | - | 8, 9 | 883 | |||
AK011869.1 | - | 1, 2 | 1506 | |||
BC026820.1 | - | 10, 11 | 1090 | 4039464 | ||
BC107273.1 | - | 10, 11 | 1003 | |||
BC107274.1 | - | 10, 11 | 1007 |