Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019505.3 | NP_062378.1 | 3 | 1034 | ||
GenBank mRNA | AF136744.1 | - | 3 | 671 | ||
AK044596.1 | - | 1770 | ||||
AK049335.1 | - | 3 | 834 | |||
AK147841.1 | - | 3 | 1035 | |||
BB635763.1 | - | 3 | 438 | |||
BC055867.1 | - | 3 | 967 |