Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prodh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019546.5 | NP_062419.2 | 5 | 650 | ||
GenBank mRNA | AF222851.1 | - | 5 | 637 | ||
AK011020.1 | - | 5 | 641 | |||
AK011039.1 | - | 5 | 650 | |||
AK050141.1 | - | 5 | 651 | |||
BC018182.1 | - | 5 | 636 | 4161894 | ||
U80019.1 | - | 5 | 648 |