Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146200.1 | NP_666312.1 | 7 | 635 | ||
GenBank mRNA | AK013160.1 | - | 7 | 662 | ||
AK076165.1 | - | 7 | 653 | |||
AK088076.1 | - | 7 | 678 | |||
AK152693.1 | - | 7 | 653 | |||
AK160078.1 | - | 7 | 655 | |||
AK166743.1 | - | 7 | 678 | |||
AK168160.1 | - | 7 | 639 | |||
AK168174.1 | - | 7 | 639 | |||
AK181388.1 | - | 2 | 106 | |||
BC024855.1 | - | 7 | 635 | 5359060 | ||
BC025032.1 | - | 7 | 656 | 4947435 |