Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284328.1 | NP_001271257.1 | 2 | 367 | ||
NM_019666.2 | NP_062640.2 | 2 | 367 | |||
NM_019796.5 | NP_062770.1 | 2 | 367 | |||
GenBank mRNA | AB035725.1 | - | 2 | 166 | ||
AF093821.1 | - | 1 | 19 | |||
AK034845.1 | - | 2 | 179 | |||
AK076026.1 | - | 2 | 159 | |||
AK077588.1 | - | 2 | 116 | |||
BC041148.1 | - | 2 | 212 | 4946652 | ||
BC050079.1 | - | 2 | 140 | 30012104 | ||
BC050215.1 | - | 1 | 484 | |||
BC055863.1 | - | 2 | 145 | |||
BC058807.1 | - | 2 | 234 | |||
BC080309.1 | - | 2 | 70 | 5716184 | ||
BC108363.1 | - | 2 | 101 | 4239809 | ||
BU505018.1 | - | 2 | 232 | |||
CV562339.1 | - | 2 | 529 |