Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gsk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019827.6 | NP_062801.1 | 1, 2 | 1605 | ||
GenBank mRNA | AB066114.1 | - | 1, 2 | 1604 | ||
AF156099.2 | - | 1, 2 | 246 | |||
AK154293.1 | - | 1, 2 | 1083 | |||
AK170965.1 | - | 1, 2 | 1052 | |||
AK171482.1 | - | 1, 2 | 1122 | |||
AK190946.1 | - | 1, 2 | 49 | |||
BC006936.1 | - | 1, 2 | 112 | 2648507 | ||
BC060743.1 | - | 1, 2 | 1532 | 4022374 |