Gene Symbol: Rabgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199059.1 | NP_001185988.1 | 6 | 732 | ||
NM_019983.2 | NP_064367.1 | 5 | 877 | |||
GenBank mRNA | AF093590.1 | - | 6 | 708 | ||
AK044568.1 | - | 5 | 897 | |||
AK146734.1 | - | 6 | 682 | |||
BC018229.1 | - | 5 | 877 | 4507477 |