Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020009.2 | NP_064393.2 | 14, 15 | 2446 | ||
GenBank mRNA | AF152838.1 | - | 14, 15 | 2421 | ||
AK143381.1 | - | 11, 12 | 1857 | |||
AK172584.1 | - | 14, 15 | 2437 | |||
BC112904.1 | - | 14, 15 | 2404 | 6465126 |