Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019993.3 | NP_064377.2 | 6 | 1097 | ||
GenBank mRNA | AB296386.1 | - | 6 | 1042 | ||
AF170919.1 | - | 6 | 842 | |||
AK132336.1 | - | 6 | 882 | |||
AK149589.1 | - | 6 | 883 | |||
AK150093.1 | - | 6 | 840 | |||
AK154914.1 | - | 6 | 1081 | |||
AK162354.1 | - | 6 | 881 | |||
AK165730.1 | - | 6 | 1097 | |||
AK168261.1 | - | 6 | 1075 | |||
AK168309.1 | - | 6 | 1075 | |||
AK168878.1 | - | 6 | 1046 | |||
AK177761.1 | - | 336 | ||||
AK179746.1 | - | 1 | 48 | |||
AK180861.1 | - | 1 | 48 | |||
AK184027.1 | - | 1 | 48 | |||
AK188805.1 | - | 1 | 48 | |||
AK199575.1 | - | 1 | 48 | |||
AK203750.1 | - | 1 | 48 | |||
AK211951.1 | - | 1 | 48 | |||
BC003297.1 | - | 6 | 874 | 3583976 |