Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019993.3 | NP_064377.2 | 11 | 1728 | ||
GenBank mRNA | AB296386.1 | - | 11 | 1673 | ||
AF170919.1 | - | 11 | 1473 | |||
AK010526.1 | - | 3 | 281 | |||
AK132336.1 | - | 11 | 1513 | |||
AK149589.1 | - | 11 | 1514 | |||
AK150093.1 | - | 11 | 1471 | |||
AK154914.1 | - | 11 | 1712 | |||
AK162354.1 | - | 11 | 1512 | |||
AK165730.1 | - | 11 | 1728 | |||
AK168261.1 | - | 11 | 1706 | |||
AK168309.1 | - | 11 | 1706 | |||
AK168878.1 | - | 11 | 1677 | |||
AK200576.1 | - | 2 | 86 | |||
AK207682.1 | - | 2 | 86 | |||
AK217939.1 | - | 2 | 79 | |||
BC003297.1 | - | 11 | 1505 | 3583976 |