Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020277.2 | NP_064673.2 | 6, 7 | 806 | ||
GenBank mRNA | AB039952.1 | - | 5, 6 | 723 | ||
AF228681.1 | - | 6, 7 | 806 | |||
AJ271092.2 | - | 5, 6 | 719 | |||
AK035197.1 | - | 2, 3 | 336 | |||
AK044908.1 | - | 2, 3 | 421 | |||
AY280364.1 | - | 6, 7 | 770 | |||
AY280365.1 | - | 6, 7 | 770 | |||
BC133712.1 | - | 5, 6 | 723 | 40143371 |