Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020560.2 | NP_065585.1 | 3, 4 | 691 | ||
GenBank mRNA | AK005110.1 | - | 3, 4 | 650 | ||
AK011825.1 | - | 3, 4 | 647 | |||
AK078073.1 | - | 3, 4 | 634 | |||
AK161913.1 | - | 3, 4 | 634 | |||
BC006764.1 | - | 3, 4 | 586 | |||
BC018475.1 | - | 3, 4 | 590 | 3492633 | ||
BC027430.1 | - | 3, 4 | 595 | 4187438 | ||
Z46966.1 | - | 3, 4 | 610 |