Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020593.2 | NP_065618.1 | 10 | 1278 | ||
NM_212433.1 | NP_997598.1 | 10 | 1278 | |||
GenBank mRNA | AF233226.1 | - | 10 | 1217 | ||
AK004544.2 | - | 10 | 1230 | |||
AK006392.1 | - | 3 | 390 | |||
AK009024.1 | - | 10 | 1217 | |||
AK019550.1 | - | 10 | 1218 | |||
AK028048.1 | - | 10 | 1240 | |||
AK029594.1 | - | 3 | 397 | |||
AK031185.1 | - | 10 | 1215 | |||
AK031428.1 | - | 10 | 1238 | |||
AK049870.1 | - | 10 | 1236 | |||
AK050195.1 | - | 10 | 1230 | |||
AK150097.1 | - | 9 | 1114 | |||
AK151130.1 | - | 10 | 1229 | |||
AK151857.1 | - | 10 | 1229 | |||
AK152780.1 | - | 10 | 1229 | |||
AK167075.1 | - | 10 | 1216 | |||
AK176959.1 | - | 3 | 313 | |||
BC010212.1 | - | 10 | 1179 | |||
BC050758.1 | - | 10 | 1231 | |||
BC058253.1 | - | 10 | 1235 | 6465107 |