Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020593.2 | NP_065618.1 | 5 | 670 | ||
NM_212433.1 | NP_997598.1 | 5 | 670 | |||
GenBank mRNA | AF233226.1 | - | 5 | 609 | ||
AK004544.2 | - | 5 | 622 | |||
AK009024.1 | - | 5 | 609 | |||
AK019550.1 | - | 5 | 610 | |||
AK028048.1 | - | 5 | 632 | |||
AK031185.1 | - | 5 | 609 | |||
AK031428.1 | - | 5 | 630 | |||
AK049870.1 | - | 5 | 628 | |||
AK050195.1 | - | 5 | 622 | |||
AK085686.1 | - | 5 | 625 | |||
AK151130.1 | - | 5 | 621 | |||
AK151857.1 | - | 5 | 621 | |||
AK152780.1 | - | 5 | 621 | |||
AK167075.1 | - | 5 | 608 | |||
BC010212.1 | - | 5 | 571 | |||
BC050758.1 | - | 5 | 623 | |||
BC058253.1 | - | 5 | 627 | 6465107 |