Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Angptl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020581.2 | NP_065606.2 | 1 | 495 | ||
GenBank mRNA | AB054540.1 | - | 1 | 381 | ||
AF123261.1 | - | 1 | 426 | |||
AF169313.1 | - | 1 | 358 | |||
AF278699.1 | - | 1 | 430 | |||
AK009161.1 | - | 1 | 201 | |||
AK014564.1 | - | 496 | ||||
AK132761.1 | - | 1 | 464 | |||
AK134419.1 | - | 1 | 464 | |||
BC006611.1 | - | 1 | 447 | 3966729 | ||
BC021343.1 | - | 1 | 461 | 4194173 | ||
BC025797.1 | - | 1 | 431 | 5137159 | ||
KJ650202.1 | - | 1 | 312 |