Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021346.2 | NP_067321.2 | 2, 3 | 725 | ||
NM_207671.4 | NP_997554.2 | 2, 3 | 725 | |||
GenBank mRNA | AF159455.1 | - | 1, 2 | 134 | ||
AF227194.1 | - | 1, 2 | 134 | |||
AW457229.1 | - | 2, 3 | 371 | |||
BC150730.1 | - | 1, 2 | 56 | 9087641 |