Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135727.2 | NP_001129199.1 | 8 | 1297 | ||
NM_001135728.2 | NP_001129200.1 | 3 | 299 | |||
NM_021389.6 | NP_067364.2 | 7 | 862 | |||
GenBank mRNA | AK004636.1 | - | 7 | 862 | ||
AK018032.1 | - | 7 | 878 | |||
AK020782.1 | - | 7 | 839 | |||
AK028299.1 | - | 3 | 300 | |||
AK030839.1 | - | 7 | 863 | |||
AK038540.1 | - | 10 | 1161 | |||
AK049182.1 | - | 8 | 1298 | |||
AK080930.1 | - | 8 | 1162 | |||
AK161735.1 | - | 3 | 380 | |||
AK172022.1 | - | 8 | 1278 | |||
BC034646.1 | - | 7 | 866 |