Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trem3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021407.3 | NP_067382.1 | 3, 4 | 621 | ||
GenBank mRNA | AF241220.1 | - | 3, 4 | 573 | ||
AK079407.1 | - | 3, 4 | 546 | |||
BC127035.1 | - | 3, 4 | 556 | 40049608 | ||
BC127036.1 | - | 3, 4 | 556 | 40049609 | ||
BY594056.1 | - | 1, 2 | 51 |