Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164197.1 | NP_001157669.1 | 1, 2 | 67 | ||
NM_021412.2 | NP_067387.1 | 1, 2 | 202 | |||
GenBank mRNA | AF153199.1 | - | 1, 2 | 154 | ||
AF155221.1 | - | 1, 2 | 182 | |||
AF162446.1 | - | 1, 2 | 182 | |||
AK079924.1 | - | 1, 2 | 68 | |||
AK080125.1 | - | 1, 2 | 62 | |||
AK136517.1 | - | 1, 2 | 56 | |||
AK154863.1 | - | 1, 2 | 195 | |||
BC112966.2 | - | 1, 2 | 174 | 40039023 |