Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021715.1 | NP_068361.1 | 1 | 1348 | ||
GenBank mRNA | AB040710.1 | - | 1 | 1348 | ||
AB046929.1 | - | 1 | 1348 | |||
AK011202.1 | - | 1 | 1360 | |||
BC019204.1 | - | 1 | 1359 | 5041967 |