Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cables1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146287.1 | NP_001139759.1 | 10 | 2203 | ||
NM_022021.2 | NP_071304.2 | 9 | 2125 | |||
GenBank mRNA | AF133208.1 | - | 9 | 2124 | ||
AF328140.1 | - | 9 | 1810 | |||
AK156273.1 | - | 10 | 2259 | |||
AK158396.1 | - | 10 | 1933 | |||
BC043661.1 | - | 9 | 1327 | |||
BC145142.1 | - | 9 | 2109 | 9053693 | ||
BC150714.1 | - | 9 | 2109 | 9087625 | ||
CX202925.1 | - | 3 | 301 |