Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cables1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146287.1 | NP_001139759.1 | 5 | 1550 | ||
NM_022021.2 | NP_071304.2 | 4 | 1472 | |||
GenBank mRNA | AF133208.1 | - | 4 | 1471 | ||
AF328140.1 | - | 4 | 1157 | |||
AK156273.1 | - | 5 | 1606 | |||
AK158396.1 | - | 5 | 1280 | |||
BC043661.1 | - | 4 | 674 | |||
BC145142.1 | - | 4 | 1456 | 9053693 | ||
BC150714.1 | - | 4 | 1456 | 9087625 | ||
BQ917292.1 | - | 2 | 157 | |||
CX213994.1 | - | 4 | 533 |