Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199113.1 | NP_001186042.1 | 10 | 1086 | ||
NM_001199114.1 | NP_001186043.1 | 9 | 925 | |||
NM_001199115.1 | NP_001186044.1 | 9 | 1025 | |||
NM_001199116.1 | NP_001186045.1 | 9 | 985 | |||
NM_022880.3 | NP_075018.1 | 8 | 1026 | |||
GenBank mRNA | AA407561.1 | - | 3 | 192 | ||
AF131212.1 | - | 8 | 1027 | |||
AF305501.1 | - | 9 | 996 | |||
AK004756.1 | - | 9 | 971 | |||
AK050089.1 | - | 9 | 1010 | |||
AK146265.1 | - | 9 | 926 | |||
AK149483.1 | - | 9 | 941 | |||
AK170463.1 | - | 9 | 957 | |||
BC004828.1 | - | 10 | 1049 | 2631728 | ||
BC006812.1 | - | 8 | 957 | 3155188 | ||
EU180577.1 | - | 8 | 901 |